ENAH31551625 Enamine

5-(trifluoromethyl)pyrimidin-2-ol

  • MDL number MFCD10696959
  • Cas Number : 83767-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94953
Availability details for
Quantity
  • Availability
  • Add to Cart