ENAH31551626 Enamine

2-acetyl-6-nitrobenzoic acid

  • MDL number MFCD18432391
  • Cas Number : 13619-70-0
  • Molecular Weight : 209.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-94973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94973
Availability details for
Quantity
  • Availability
  • Add to Cart