ENAH31551628 Enamine

1,4-dibromo-2-(2-chloroethoxy)-5-methoxybenzene

  • MDL number MFCD32069459
  • Cas Number : 281678-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97716
Availability details for
Quantity
  • Availability
  • Add to Cart