ENAH31551648 Enamine

3,6-dichloro-4-methoxypyridazine

  • MDL number MFCD00477252
  • Cas Number : 70952-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-209402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209402
Availability details for
Quantity
  • Availability
  • Add to Cart