ENAH3155168A Enamine

2-(4-bromo-3-fluorophenyl)-1,3-dioxolane

  • MDL number MFCD30631568
  • Cas Number : 940314-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-267550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267550
Availability details for
Quantity
  • Availability
  • Add to Cart