ENAH315516B8 Enamine

3-ethenyloxetan-3-ol

  • MDL number MFCD14586454
  • Cas Number : 1207175-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-316574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-316574
Availability details for
Quantity
  • Availability
  • Add to Cart