ENAH315516B9 Enamine

{2-hydroxy-3-[4-(2-methoxyethyl)phenoxy]propyl}(propan-2-yl)amine

  • MDL number MFCD00599534

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317058
Availability details for
Quantity
  • Availability
  • Add to Cart