ENAH315516E6 Enamine

3-fluorofuran

  • MDL number MFCD15143724
  • Cas Number : 2105-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-716297

Safety Information

signalword Danger
hcodes F,Xn
MSDS https://www.enaminestore.com/catalog/EN300-716297
pictograms GHS02,GHS07,GHS08
hstatements EUH019,H225,H302,H312,H332,H373
Availability details for
Quantity
  • Availability
  • Add to Cart