ENAH315516F3 Enamine

3-hydroxy-3a-methyl-hexahydro-1lambda6-pyrrolo[1,2-b][1,2]thiazole-1,1-dione

  • MDL number MFCD31794977
  • Cas Number : 2167602-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-760915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-760915
Availability details for
Quantity
  • Availability
  • Add to Cart