ENAH315516F8 Enamine

{1-oxaspiro[4.6]undecan-2-yl}methanesulfonamide

  • MDL number MFCD31745998
  • Cas Number : 2287301-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-781743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-781743
Availability details for
Quantity
  • Availability
  • Add to Cart