ENAH315516F9 Enamine

1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-1,2,3,6-tetrahydropyridine-4-carboxylic acid

  • MDL number MFCD02682418
  • Cas Number : 1185295-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-814222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-814222
Availability details for
Quantity
  • Availability
  • Add to Cart