ENAH31551700 Enamine

[(5-cyclopropyl-1,2-oxazol-3-yl)methyl](ethyl)amine

  • MDL number MFCD17216015
  • Cas Number : 1248919-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-871848

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-871848
pictograms GHS05,GHS07
hstatements H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart