ENAH31551708 Enamine

5-bromopyrimidine-2-carbohydrazide

  • MDL number MFCD00507173
  • Cas Number : 87362-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1223177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1223177
Availability details for
Quantity
  • Availability
  • Add to Cart