ENAH3155171E Enamine

6-methoxypyrimidine-2,4-diamine

  • MDL number MFCD02091088
  • Cas Number : 3270-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-169
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1242939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1242939
Availability details for
Quantity
  • Availability
  • Add to Cart