ENAH31551728 Enamine

N-ethyl-N-methylacetamide

  • MDL number MFCD20357058
  • Cas Number : 38806-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1251149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1251149
Availability details for
Quantity
  • Availability
  • Add to Cart