ENAH31551958 Enamine

8-(difluoromethoxy)quinolin-4-ol

  • MDL number MFCD18416100
  • Cas Number : 629644-63-9
  • Molecular Weight : 211.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6364600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6364600
Availability details for
Quantity
  • Availability
  • Add to Cart