ENAH315519F5 Enamine

tert-butyl N-{9-oxobicyclo[3.3.1]nonan-2-yl}carbamate

  • MDL number MFCD31746156
  • Cas Number : 2287334-10-3
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6495026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6495026
Availability details for
Quantity
  • Availability
  • Add to Cart