ENAH31551A1D Enamine

tert-butyl 5-(tributylstannyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD31745889
  • Molecular Weight : 458.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6731093

Safety Information

signalword Danger
pcodes P260,P264,P270,P273,P280,P301+P310,P302+P352,P305+P351+P338,P312,P314,P330,P332+P313,P337+P313,P362,P363,P391,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6731093
pictograms GHS06,GHS08,GHS09
hstatements H301,H312,H315,H319,H372,H400,H410
Availability details for
Quantity
  • Availability
  • Add to Cart