ENAH31551A27 Enamine

lithium(1+) ion 2-(difluoromethyl)-4-methoxybenzene-1-sulfinate

  • MDL number MFCD31794790
  • Cas Number : 2287335-46-8
  • Molecular Weight : 228.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6732320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6732320
Availability details for
Quantity
  • Availability
  • Add to Cart