ENAH31551A43 Enamine

(3aR,5R,6aS)-2-{[(9H-fluoren-9-yl)methoxy]carbonyl}-octahydrocyclopenta[c]pyrrole-5-carboxylic acid

  • MDL number MFCD31746282
  • Cas Number : 2287247-89-4
  • Molecular Weight : 377.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6733529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6733529
Availability details for
Quantity
  • Availability
  • Add to Cart