ENAH31551A44 Enamine

(1r,3r)-3-(2-fluorophenyl)cyclobutan-1-ol

  • MDL number MFCD31746284
  • Cas Number : 2287345-16-6
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6733583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6733583
Availability details for
Quantity
  • Availability
  • Add to Cart