ENAH31551A59 Enamine

5H,6H,7H,8H-imidazo[1,2-b]pyridazine

  • MDL number MFCD21336093
  • Cas Number : 54760-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6496556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6496556
Availability details for
Quantity
  • Availability
  • Add to Cart