ENAH31551A65 Enamine

tert-butyl 3,6-dimethyl-1,4-diazepane-1-carboxylate

  • MDL number MFCD31736977
  • Cas Number : 2260932-07-6
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6497434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6497434
Availability details for
Quantity
  • Availability
  • Add to Cart