ENAH31551A7B Enamine

5-(4-fluorophenyl)-1,4-oxazepane

  • MDL number MFCD31737383
  • Cas Number : 1507555-47-6
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6498605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6498605
Availability details for
Quantity
  • Availability
  • Add to Cart