ENAH31551A7F Enamine

1-(benzylsulfanyl)-3-bromo-2-nitrobenzene

  • MDL number MFCD31737384
  • Cas Number : 2260933-25-1
  • Molecular Weight : 324.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6498729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6498729
Availability details for
Quantity
  • Availability
  • Add to Cart