ENAH31551A9A Enamine

butyl 3-methylquinoline-4-carboxylate

  • MDL number MFCD18448803
  • Cas Number : 19764-43-3
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6499708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6499708
Availability details for
Quantity
  • Availability
  • Add to Cart