ENAH31551AA4 Enamine

{2-oxatricyclo[3.3.1.1,3,7]decan-1-yl}methanamine

  • Cas Number : 106237-15-4
  • Molecular Weight : 167.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6503663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6503663
Availability details for
Quantity
  • Availability
  • Add to Cart