ENAH31551B1C Enamine

3-methoxy-6-methylpyridazine

  • MDL number MFCD09751662
  • Cas Number : 17644-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729966
Availability details for
Quantity
  • Availability
  • Add to Cart