ENAH31551B5F Enamine

2-nitro-4-(propan-2-yl)phenol

  • MDL number MFCD00094157
  • Cas Number : 1576-10-9
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-55
Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6731208

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P273,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6731208
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335,H412
Availability details for
Quantity
  • Availability
  • Add to Cart