ENAH31551B7E Enamine

(1R)-1-(4-chlorophenyl)ethane-1,2-diol

  • MDL number MFCD19689566
  • Cas Number : 152142-03-5
  • Molecular Weight : 172.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6731373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6731373
Availability details for
Quantity
  • Availability
  • Add to Cart