ENAH31551B8B Enamine

3-(oxiran-2-yl)propane-1-sulfonamide

  • MDL number MFCD31736733
  • Cas Number : 2260935-82-6
  • Molecular Weight : 165.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6731452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6731452
Availability details for
Quantity
  • Availability
  • Add to Cart