ENAH31551B92 Enamine

ethyl 4-oxopent-2-ynoate

  • MDL number MFCD18971726
  • Cas Number : 54966-49-3
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6731473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6731473
Availability details for
Quantity
  • Availability
  • Add to Cart