ENAH31551BD5 Enamine

3-(2,2,2-trifluoroethoxy)-1H-pyrazol-4-amine

  • MDL number MFCD31745922
  • Cas Number : 2287312-31-4
  • Molecular Weight : 181.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6731829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6731829
Availability details for
Quantity
  • Availability
  • Add to Cart