ENAH31551BDC Enamine

2-(1-{[(9H-fluoren-9-yl)methoxy]carbonyl}piperidin-3-yl)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD31737327
  • Cas Number : 2260933-43-3
  • Molecular Weight : 448.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6731860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6731860
Availability details for
Quantity
  • Availability
  • Add to Cart