ENAH31551BE7 Enamine

ethyl 2-(4-hydroxypiperidin-4-yl)-2-methylpropanoate hydrochloride

  • MDL number MFCD31736787
  • Cas Number : 2260937-49-1
  • Molecular Weight : 251.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6731950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6731950
Availability details for
Quantity
  • Availability
  • Add to Cart