ENAH31551C07 Enamine

(3R,4S)-4-methyloxolan-3-ol

  • MDL number MFCD30491115
  • Cas Number : 1932478-75-5
  • Molecular Weight : 102.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6732250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6732250
Availability details for
Quantity
  • Availability
  • Add to Cart