ENAH31551C08 Enamine

4-hydrazinyl-1-(4-methylbenzenesulfonyl)azepane hydrochloride

  • MDL number MFCD23701348
  • Cas Number : 1429056-19-8
  • Molecular Weight : 319.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6732255

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6732255
Availability details for
Quantity
  • Availability
  • Add to Cart