ENAH31551C7F Enamine

2-(1-{[(9H-fluoren-9-yl)methoxy]carbonyl}piperidin-4-yl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD31736739
  • Cas Number : 2260930-99-0
  • Molecular Weight : 418.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6732809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6732809
Availability details for
Quantity
  • Availability
  • Add to Cart