ENAH31551C88 Enamine

1-amino-3-(2-cyanoethyl)thiourea

  • MDL number MFCD19202287
  • Cas Number : 116974-14-2
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6732872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6732872
Availability details for
Quantity
  • Availability
  • Add to Cart