ENAH31551CA6 Enamine

2-bromo-5-[(Z)-2-bromo-2-nitroethenyl]furan

  • Cas Number : 189935-72-6
  • Molecular Weight : 296.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6733026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6733026
Availability details for
Quantity
  • Availability
  • Add to Cart