ENAH31551CA7 Enamine

2-[(2S)-1-[(tert-butoxy)carbonyl]-2,3-dihydro-1H-indol-2-yl]acetic acid

  • MDL number MFCD31745963
  • Cas Number : 2287239-24-9
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6733031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6733031
Availability details for
Quantity
  • Availability
  • Add to Cart