ENAH31551CAA Enamine

(3-methoxy-5-methyl-1,2-oxazol-4-yl)methanol

  • MDL number MFCD30181922
  • Cas Number : 75989-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6481256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6481256
Availability details for
Quantity
  • Availability
  • Add to Cart