ENAH31551CE9 Enamine

tert-butyl (2E)-3-[4-(benzyloxy)phenyl]prop-2-enoate

  • Cas Number : 691905-29-0
  • Molecular Weight : 310.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6733256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6733256
Availability details for
Quantity
  • Availability
  • Add to Cart