ENAH31551CF8 Enamine

6-ethynylspiro[3.3]heptan-2-one

  • MDL number MFCD31736837
  • Cas Number : 2122322-27-2
  • Molecular Weight : 134.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6733306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6733306
Availability details for
Quantity
  • Availability
  • Add to Cart