ENAH31551D04 Enamine

3-(3,3-dimethoxycyclobutyl)pyrrolidine

  • MDL number MFCD31746154
  • Cas Number : 2287334-04-5
  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6494579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6494579
Availability details for
Quantity
  • Availability
  • Add to Cart