ENAH31551D17 Enamine

(2S)-6-(1-aminocyclopropyl)-7-fluoro-2-methyl-10-oxo-4-oxa-1-azatricyclo[7.3.1.0,5,13]trideca-5(13),6,8,11-tetraene-11-carboxylic acid; methanesulfonic acid

  • MDL number MFCD00913262
  • Cas Number : 163680-77-1
  • Molecular Weight : 414.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3S)-10-(1-aminocyclopropyl)-9-fluoro-3-methyl-7-oxo-2,3-dihydro-7H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid methanesulfonate
Inchi Code 1S/C16H15FN2O4.CH4O3S/c1-7-6-23-14-11(16(18)2-3-16)10(17)4-8-12(14)19(7)5-9(13(8)20)15(21)22;1-5(2,3)4/h4-5,7H,2-3,6,18H2,1H3,(H,21,22);1H3,(H,2,3,4)/t7-;/m0./s1
Inchi Key UDHGFPATQWQARM-FJXQXJEOSA-N

Description

Purity 95
Salt Data CS(=O)(=O)O
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6733488

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6733488
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart