ENAH31551D18 Enamine

5-chloro-N-{[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methyl}thiophene-2-carboxamide

  • MDL number MFCD11974010
  • Cas Number : 366789-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228-229
Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6733490

Safety Information

pcodes P273,P391,P501
MSDS https://www.enaminestore.com/catalog/EN300-6733490
pictograms GHS09
hstatements H411
Availability details for
Quantity
  • Availability
  • Add to Cart