ENAH31551D1A Enamine

(2S,3R,4R,5S,6R)-2-(3-{[5-(4-fluorophenyl)thiophen-2-yl]methyl}-4-methylphenyl)-6-(hydroxymethyl)oxane-3,4,5-triol

  • MDL number MFCD18251436

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6733492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6733492
Availability details for
Quantity
  • Availability
  • Add to Cart