ENAH31551D41 Enamine

4,4-dimethylpentanimidamide hydrochloride

  • MDL number MFCD28895394
  • Cas Number : 64285-97-8
  • Molecular Weight : 164.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6733700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6733700
Availability details for
Quantity
  • Availability
  • Add to Cart