ENAH31551D89 Enamine

5-bromo-1-hydroxy-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD31794794
  • Cas Number : 2287343-14-8
  • Molecular Weight : 234.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6734071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734071
Availability details for
Quantity
  • Availability
  • Add to Cart